2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-(2-Phenylthiazol-4-yl)acetic acid
H54805.MD
H54805.03
H54805.MDEA
0
EUR
H54805.MD
H54805.03
2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid 97%
2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid
Purity: 97%
CAS No.: 16441-28-4
Molecular Formula: C11H9NO2S
Formula Weight: 219.26
MDL No.: MFCD01312954
Formula:
C₁₁H₉NO₂S Storage Temperature: Ambient |
MDL Number:
MFCD01312954 CAS Number: 16441-28-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...