1-Amino-4-cyclopentylpiperazine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Cyclopentylpiperazin-1-amine

H54831.MD H54831.03 H54831.06
H54831.MDEA 112 EUR
H54831.MD H54831.03 H54831.06
1-Amino-4-cyclopentylpiperazine 97%
1-Amino-4-cyclopentylpiperazine

Purity: 97% CAS No.: 61379-64-4 Molecular Formula: C9H19N3 Formula Weight: 169.27 MDL No.: MFCD00190598

Formula: C₉H₁₉N₃
Storage Temperature: Ambient
MDL Number: MFCD00190598
CAS Number: 61379-64-4
EINECS: 262-742-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR