2-(4-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H54920.MD
H54920.03
H54920.MDEA
0
EUR
H54920.MD
H54920.03
2-(4-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione 97%
2-(4-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione
Purity: 97%
Molecular Formula: C20H11N3O2
Formula Weight: 325.33
MDL No.: MFCD20265373
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...