3,5-Di-tert-butylsalicylic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5-Bis-tert-butylsalicylic acid, 3,5-Di-tert-butyl-2-hydroxybenzoic Acid

H55102.14 H55102.06
H55102.14EA 126 EUR
H55102.14 H55102.06
3,5-Di-tert-butylsalicylic acid 97%
3,5-Di-tert-butylsalicylic acid

Purity: 97% CAS No.: 19715-19-6 Molecular Formula: C15H22O3 Formula Weight: 250.34 Flash Point: 179°(354°F) Melting Point: 157-162° MDL No.: MFCD00059058

Formula: C₁₅H₂₂O₃
MW: 250,34 g/mol
Melting Pt: 157…162 °C
Flash Pt: 179 °C (354 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059058
CAS Number: 19715-19-6
EINECS: 243-246-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR