1-(4-Aminophenyl)piperazine 97%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Piperazin-1-ylaniline

H55224.MD H55224.03 H55224.06
H55224.MDEA 0 EUR
H55224.MD H55224.03 H55224.06
1-(4-Aminophenyl)piperazine 97%
1-(4-Aminophenyl)piperazine

Purity: 97% CAS No.: 67455-41-8 Molecular Formula: C10H15N3 Formula Weight: 177.25 Physical Form: Powder Appearance: White to purple Melting Point: 119-123° MDL No.: MFCD01365906

Formula: C₁₀H₁₅N₃
MW: 177,25 g/mol
Melting Pt: 119…123 °C
Storage Temperature: Ambient
MDL Number: MFCD01365906
CAS Number: 67455-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR