4-tert-Butyldimethylsiloxy-1-butanol 97%
Supplier: Thermo Scientific
H55285.06
H55285.14
H55285.06EA
52.8
EUR
H55285.06
H55285.14
4-tert-Butyldimethylsiloxy-1-butanol 97%
4-tert-Butyldimethylsiloxy-1-butanol
Purity: 97%
CAS No.: 87184-99-4
Molecular Formula: C10H24O2Si
Formula Weight: 204.38
Flash Point: 105°(221°F)
Refractive Index: 1.4390
Boiling Point: 63-66°/0.3mm
MDL No.: MFCD04974070
Formula:
C₁₀H₂₄O₂Si Boiling Pt: 63…66 °C (0,3 mmHg) Density: 0,888 Flash Pt: 105 °C (221 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04974070 CAS Number: 87184-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...