8-(Tosylamino)quinoline 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H55386.06
H55386.14
H55386.06EA
0
EUR
H55386.06
H55386.14
8-(Tosylamino)quinoline 97%
8-(Tosylamino)quinoline
Purity: 97%
CAS No.: 10304-39-9
Molecular Formula: C16H14N2O2S
Formula Weight: 298.36
Physical Form: Solid
Appearance: White to pale brown
Melting Point: 154-158°
Beilstein Reference: 264159
MDL No.: MFCD00168990
Formula:
C₁₆H₁₄N₂O₂S Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD00168990 CAS Number: 10304-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...