Benzyl(4-methoxybenzyl)amine 97%

Supplier: Thermo Fisher Scientific
Danger

H55633.MD H55633.03
H55633.MDEA 0 EUR
H55633.MD H55633.03
Benzyl(4-methoxybenzyl)amine 97%
Benzyl(4-methoxybenzyl)amine

Purity: 97% CAS No.: 14429-02-8 Molecular Formula: C15H17NO Formula Weight: 227.31 UN No.: UN3259 MDL No.: MFCD01002920

Formula: C₁₅H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD01002920
CAS Number: 14429-02-8
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR