Tris(8-quinolinolato)aluminum 99%

Supplier: Thermo Scientific
Warning

Synonyms: 8-Quinolinol Aluminum Complex, Alq3, Aluminum Tris(8-hydroxyquinolinate), Aluminum triquinolin-8-olate

H55656.03 H55656.06 H55656.14
H55656.03EA 31.5 EUR
H55656.03 H55656.06 H55656.14
Tris(8-quinolinolato)aluminum 99%
Tris(8-quinolinolato)aluminum

Purity: 99% CAS No.: 2085-33-8 Molecular Formula: C27H18AlN3O3 Formula Weight: 459.44 Physical Form: Powder Appearance: Yellow to green Melting Point: 300° MDL No.: MFCD00191693

Formula: C₂₇H₁₈AlN₃O₃
MW: 459,44 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191693
CAS Number: 2085-33-8
EINECS: 218-227-0
Merck Index: 14,370

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR