1-Acetyladamantane 99%

Supplier: Thermo Scientific

Synonyms: 1-Adamantyl Methyl Ketone, Methyl 1-Adamantyl Ketone

H55889.03 H55889.06
H55889.03EA 46.7 EUR
H55889.03 H55889.06
1-Acetyladamantane 99%
1-Acetyladamantane

Purity: 99% CAS No.: 1660-04-4 Molecular Formula: C12H18O Formula Weight: 178.27 Flash Point: 108°(226°F) Melting Point: 53-55° MDL No.: MFCD00074739

Formula: C₁₂H₁₈O
MW: 178,28 g/mol
Melting Pt: 53…55 °C
Flash Pt: 108 °C (226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00074739
CAS Number: 1660-04-4
EINECS: 216-761-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR