N-Boc-L-β-proline ethyl ester 97%
Supplier: Thermo Scientific
|
Warning
|
H57034.MD
H57034.03
H57034.MDEA
31.5
EUR
H57034.MD
H57034.03
N-Boc-L-β-proline ethyl ester 97%
N-Boc-L-beta-proline ethyl ester
Purity: 97%
Molecular Formula: C12H21NO4
Formula Weight: 243.30
Storage Temperature: 2°C to 8°C
MDL No.: MFCD06204615
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...