(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate 95%

Supplier: Thermo Scientific
Warning

Synonyms: (S)-1-Boc-2-pyrrolidinecarbonitrile, (S)-1-(tert-Butoxycarbonyl)-2-cyanopyrrolidine, (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinecarbonitrile, (S)-1-Boc-2-cyanopyrrolidine, tert-Butyl (S)-2-Cyano-1-pyrrolidinecarboxylate, (S)-tert-Butyl 2-cyanopyrrolidine-1-carboxylate, (S)-2-Cyano-1-pyrrolidinecarboxyic acid tert-Butyl ester

H57150.MD H57150.03
H57150.MDEA 95.6 EUR
H57150.MD H57150.03
(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate 95%
(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate

Purity: 95% CAS No.: 228244-04-0 Molecular Formula: C10H16N2O2 Formula Weight: 196.25 Flash Point: 110°(230°F) Melting Point: 33-36° Optical Rotation: +105° (c=1 in chloroform) MDL No.: MFCD01861224

Formula: C₁₀H₁₆N₂O₂
MW: 196,25 g/mol
Melting Pt: 33…36 °C
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01861224
CAS Number: 228244-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR