N-Boc-D-β-proline ethyl ester 97%
Supplier: Thermo Scientific
|
Warning
|
H57390.MD
H57390.03
H57390.MDEA
0
EUR
H57390.MD
H57390.03
N-Boc-D-β-proline ethyl ester 97%
N-Boc-D-beta-proline ethyl ester
Purity: 97%
CAS No.: 1314419-65-2
Molecular Formula: C12H21NO4
Formula Weight: 243.30
Storage Temperature: 2°C to 8°C
MDL No.: MFCD19442798
Formula:
C₁₂H₂₁NO₄ MW: 243,30 g/mol |
MDL Number:
MFCD19442798 CAS Number: 1314419-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...