4-Methylpiperazin-2-one 95%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Methyl-3-oxopiperazine

H57633.MD H57633.03
H57633.MDEA 0 EUR
H57633.MD H57633.03
4-Methylpiperazin-2-one 95%
4-Methylpiperazin-2-one

Purity: 95% CAS No.: 34770-60-0 Molecular Formula: C5H10N2O Formula Weight: 114.15 Melting Point: 84-90° MDL No.: MFCD11870124

Formula: C₅H₁₀N₂O
MW: 114,15 g/mol
Melting Pt: 84…90 °C
Storage Temperature: Ambient
MDL Number: MFCD11870124
CAS Number: 34770-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR