(S)-1-Boc-(3-hydroxymethyl)pyrrolidine ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: tert-butyl (3S)-3-(hydroxymethyl)pyrrolidine-1-carboxylate, (S)-1-Boc-(3-Hydroxymethyl)pyrrolidine

H57753.MD H57753.03
H57753.MDEA 74.4 EUR
H57753.MD H57753.03
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine ≥97%
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine

Purity: 97+% CAS No.: 199174-24-8 Molecular Formula: C10H19NO3 Formula Weight: 201.27 Boiling Point: 285-289° MDL No.: MFCD03094726

Formula: C₁₀H₁₉NO₃
MW: 201,27 g/mol
Boiling Pt: 285…289 °C
Density: 1,084
Storage Temperature: Ambient
MDL Number: MFCD03094726
CAS Number: 199174-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR