2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Methyl-N-(oxetan-3-ylidene)propane-2-sulphinamide

H57776.MD H57776.ME
H57776.MDEA 0 EUR
H57776.MD H57776.ME
2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide 95%
2-Methyl-N-(3-oxetanylidene)propane-2-sulphinamide

Purity: 95% CAS No.: 1158098-73-7 Molecular Formula: C7H13NO2S Formula Weight: 175.25 MDL No.: MFCD14702522

Formula: C₇H₁₃NO₂S
MW: 175,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14702522
CAS Number: 1158098-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR