4-(2-Furyl)-2-methylthiazole 97%

Supplier: Thermo Scientific
Warning

H58115.03 H58115.06
H58115.03EA 207 EUR
H58115.03 H58115.06
4-(2-Furyl)-2-methylthiazole 97%
4-(2-Furyl)-2-methylthiazole

Purity: 97% CAS No.: 492440-71-8 Molecular Formula: C8H7NOS Formula Weight: 165.20 Boiling Point: 128° Beilstein Reference: 128400 MDL No.: MFCD22682939

Formula: C₈H₇NOS
MW: 165,20 g/mol
Boiling Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD22682939
CAS Number: 492440-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR