2,4-Dichloronicotinc acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H58239.MD
H58239.03
H58239.MDEA
0
EUR
H58239.MD
H58239.03
2,4-Dichloronicotinc acid 97%
2,4-Dichloronicotinc acid
Purity: 97%
CAS No.: 262423-77-8
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Melting Point: 166-167°
MDL No.: MFCD06796292
Formula:
C₆H₃Cl₂NO₂ Melting Pt: 166…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD06796292 CAS Number: 262423-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...