3-Chromanecarboxylic acid 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Chroman-3-carboxylic acid
H58448.03
H58448.06
H58448.03EA
104
EUR
H58448.03
H58448.06
3-Chromanecarboxylic acid 97%
3-Chromanecarboxylic acid
Purity: 97%
CAS No.: 115822-57-6
Molecular Formula: C10H10O3
Formula Weight: 178.18
Melting Point: 117-121°
MDL No.: MFCD03783554
Formula:
C₁₀H₁₀O₃ MW: 178,19 g/mol Melting Pt: 117…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD03783554 CAS Number: 115822-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...