2-(Boc-amino)-4-phenylthiazole-5-carboxamidoxime 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H58529.03
H58529.06
H58529.03EA
0
EUR
H58529.03
H58529.06
2-(Boc-amino)-4-phenylthiazole-5-carboxamidoxime 97%
2-(Boc-amino)-4-phenylthiazole-5-carboxamidoxime
Purity: 97%
Molecular Formula: C15H18N4O3S
Formula Weight: 334.39
MDL No.: MFCD22988963
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...