Thiophene-3-carboxamidoxime 97%

Supplier: Thermo Scientific
Warning

Synonyms: Thiophene-3-amidoxime

H58858.03 H58858.06
H58858.03EA 199 EUR
H58858.03 H58858.06
Thiophene-3-carboxamidoxime 97%
Thiophene-3-carboxamidoxime

Purity: 97% CAS No.: 58905-71-8 Molecular Formula: C5H6N2OS Formula Weight: 142.17 MDL No.: MFCD01313358

Formula: C₅H₆N₂OS
MW: 142,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01313358
CAS Number: 58905-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR