Boc-Tic-OH 98%
Supplier: Thermo Scientific
Synonyms:
(S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-Tic-OH, (S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
H61423.06
H61423.14
H61423.06EA
95.6
EUR
H61423.06
H61423.14
Boc-Tic-OH 98%
Boc-Tic-OH
Purity: 98%
CAS No.: 78879-20-6
Molecular Formula: C15H19NO4
Formula Weight: 277.32
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to off-white
Beilstein Reference: 4297114
MDL No.: MFCD00143845
Formula:
C₁₅H₁₉NO₄ MW: 277,32 g/mol |
MDL Number:
MFCD00143845 CAS Number: 78879-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...