3-(Bromomethyl)thiophene 97%

Supplier: Thermo Scientific
Warning

H61497.06
H61497.06EA 399 EUR
H61497.06
3-(Bromomethyl)thiophene 97%
3-(Bromomethyl)thiophene
Formula: C₅H₅BrS
MW: 177,06 g/mol
Boiling Pt: 76 °C (1 mmHg)
Density: 1,616
MDL Number: MFCD03840497
CAS Number: 34846-44-1
EINECS: 252-247-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR