Boc-DAP(Z)-OH·DCHA 98%
Supplier: Thermo Scientific
Synonyms:
(S)-3-Benzyloxycarbonylamino-2-(Boc-amino)propionic acid dicyclohexylammonium salt, Boc-Dap(Z)-OH.DCHA, Boc-Dap(Z)-OH·DCHA
H61558.03
H61558.06
H61558.03EA
83.1
EUR
H61558.03
H61558.06
Boc-DAP(Z)-OH·DCHA 98%
Boc-DAP(Z)-OH·DCHA
Purity: 98%
CAS No.: 65710-58-9
Molecular Formula: C16H22N2O6.C12H23N
Formula Weight: 385.54
Physical Form: Solid
Beilstein Reference: 4100906
MDL No.: MFCD00236880
Formula:
C₂₈H₄₅N₃O₆ MW: 519,683 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00236880 CAS Number: 65710-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...