Boc-Lys(boc)-OSu 97%
Supplier: Thermo Fisher Scientific
Synonyms:
(S)-2,5-Dioxopyrrolidin-1-yl 2,6-bis((tert-butoxycarbonyl)amino)hexanoate, Nα,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester
H62068.06
H62068.14
H62068.06EA
136
EUR
H62068.06
H62068.14
Boc-Lys(boc)-OSu 97%
Boc-Lys(boc)-OSu
Purity: 97%
CAS No.: 30189-36-7
Molecular Formula: C20H33N3O8
Formula Weight: 443.49
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Off-white
Melting Point: 184°
Beilstein Reference: 1559007
Optical Rotation: -25°(c=1 in DMF)
MDL No.: MFCD00057898
Formula:
C₂₀H₃₃N₃O₈ Melting Pt: 184 °C |
MDL Number:
MFCD00057898 CAS Number: 30189-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...