N-Boc-3-(4-pyridyl)-D-alanine 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Boc-D-4-Pal-OH

H62215.03 H62215.06
H62215.03EA 203 EUR
H62215.03 H62215.06
N-Boc-3-(4-pyridyl)-D-alanine 95%
N-Boc-3-(4-pyridyl)-D-alanine

Purity: 95% CAS No.: 37535-58-3 Molecular Formula: C13H18N2O4 Formula Weight: 266.29 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White Melting Point: 223-229° Beilstein Reference: 4195478 Optical Rotation: +4.1°(c=0.6 in acetic acid) MDL No.: MFCD00672529

Formula: C₁₃H₁₈N₂O₄
MW: 266.29 g/mol
Melting Pt: 223…229 °C
MDL Number: MFCD00672529
CAS Number: 37535-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR