4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific

Synonyms: 5-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Fluoro-2-hydroxyphenylboronic acid pinacol ester

H62252.MD H62252.03
H62252.MDEA 0 EUR
H62252.MD H62252.03
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester 96%
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C12H16BFO3 Formula Weight: 238.06 Physical Form: Solid

Formula: C₁₂H₁₆BFO₃
MW: 238,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16994248
CAS Number: 1038828-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR