Boc-Glu-NH₂ 98%
Supplier: Thermo Scientific
Synonyms:
4-(Boc-amino)-4-carbamoylbutyric acid
H62305.03
H62305.06
H62305.03EA
91.1
EUR
H62305.03
H62305.06
Boc-Glu-NH₂ 98%
Boc-Glu-NH₂
Purity: 98%
CAS No.: 18800-74-3
Molecular Formula: C10H18N2O5
Formula Weight: 246.26
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00070081
Formula:
C₁₀H₁₈N₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00070081 CAS Number: 18800-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...