4-Fluoro-2-methoxybenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific

Synonyms: 2-(4-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4-Fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 4-Fluoro-2-methoxyphenylboronic acid pinacol ester

H62391.MD H62391.03
H62391.MDEA 0 EUR
H62391.MD H62391.03
4-Fluoro-2-methoxybenzeneboronic acid pinacol ester 96%
4-Fluoro-2-methoxybenzeneboronic acid pinacol ester

Purity: 96% CAS No.: 624741-83-9 Molecular Formula: C13H18BFO3 Formula Weight: 252.09 Physical Form: Solid

Formula: C₁₃H₁₈BFO₃
MW: 252,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18732613
CAS Number: 624741-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR