Methyl (3-bromo-5-fluorophenyl)acetate 96%

Supplier: Thermo Scientific

Synonyms: Methyl 2-(3-bromo-5-fluorophenyl)acetate

H62561.03 H62561.06
H62561.03EA 254 EUR
H62561.03 H62561.06
Methyl (3-bromo-5-fluorophenyl)acetate 96%
Methyl (3-bromo-5-fluorophenyl)acetate

Purity: 96% CAS No.: 937732-38-2 Molecular Formula: C9H8BrFO2 Formula Weight: 247.06 Physical Form: Solid

Formula: C₉H₈BrFO₂
MW: 247,063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20489056
CAS Number: 937732-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR