D-(-)-Cysteine 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(S)-2-Amino-3-mercaptopropanoic acid, H-D-Cys-OH, (S)-(-)-Cysteine
H62627.03
H62627.06
H62627.03EA
118
EUR
H62627.03
H62627.06
D-(-)-Cysteine 98%
D(-)-Cysteine
Purity: 98%
CAS No.: 921-01-7
Molecular Formula: C3H7NO2S
Formula Weight: 121.16
Physical Form: Solid
Sensitivity: Air Sensitive
Melting Point: 230° dec
Beilstein Reference: 1721407
MDL No.: MFCD00066461
Formula:
C₃H₇NO₂S MW: 121.16 g/mol Melting Pt: 230 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066461 CAS Number: 921-01-7 EINECS: 213-062-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...