6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol 96%
Supplier: Thermo Fisher Scientific
H62631.MD
H62631.03
H62631.MDEA
0
EUR
H62631.MD
H62631.03
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol 96%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol
Purity: 96%
CAS No.: 269410-21-1
Molecular Formula: C16H19BO3
Formula Weight: 270.13
Physical Form: Solid
Appearance: White
MDL No.: MFCD20486767
Formula:
C₁₆H₁₉BO₃ Storage Temperature: Ambient |
MDL Number:
MFCD20486767 CAS Number: 269410-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...