4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane 96%
Supplier: Thermo Fisher Scientific
Synonyms:
(1-Naphthylmethyl)boronic acid pinacol ester
H62764.MD
H62764.03
H62764.MDEA
0
EUR
H62764.MD
H62764.03
4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane 96%
4,4,5,5-Tetramethyl-2-(naphthalen-1-ylmethyl)-1,3,2-dioxaborolane
Purity: 96%
CAS No.: 475250-57-8
Molecular Formula: C17H21BO2
Formula Weight: 268.16
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD22419278
Formula:
C₁₇H₂₁BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22419278 CAS Number: 475250-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...