H-D-Lys(Z)-OH 95%

Supplier: Thermo Fisher Scientific

Synonyms: H-D-Lys(Cbz)-OH, Nepsilon-Benzyloxycarbonyl-D-lysine, Nepsilon-Cbz-D-lysine, H-D-Lys(Z)-OH, Nepsilon-Carbobenzoxy-D-lysine

H62819.03 H62819.06 H62819.14
H62819.03EA 0 EUR
H62819.03 H62819.06 H62819.14
H-D-Lys(Z)-OH 95%
H-D-Lys(Z)-OH

Purity: 95% CAS No.: 34404-32-5 Molecular Formula: C14H20N2O4 Formula Weight: 280.32 Physical Form: Powder Appearance: White MDL No.: MFCD00063120

Formula: C₁₄H₂₀N₂O₄
MW: 280.324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063120
CAS Number: 34404-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR