3-Fluoro-5-methoxybenzeneboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific

Synonyms: 1,3,2-Dioxaborolane, 2-(3-fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-, 3-Fluoro-5-methoxyphenylboronic acid pinacol ester, 2-(3-Fluoro-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H62841.MD H62841.03
H62841.MDEA 0 EUR
H62841.MD H62841.03
3-Fluoro-5-methoxybenzeneboronic acid pinacol ester 96%
3-Fluoro-5-methoxybenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C13H18BFO3 Formula Weight: 252.09 Physical Form: Solid

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996298
CAS Number: 1416367-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR