N-Boc-2-bromo-D-phenylalanine 95%
Supplier: Thermo Fisher Scientific
Synonyms:
Boc-D-Phe(2-Br)-OH
H62848.03
H62848.06
H62848.03EA
0
EUR
H62848.03
H62848.06
N-Boc-2-bromo-D-phenylalanine 95%
N-Boc-2-bromo-D-phenylalanine
Purity: 95%
CAS No.: 261360-76-3
Molecular Formula: C14H18BrNO4
Formula Weight: 344.20
Physical Form: Powder
Appearance: White
MDL No.: MFCD01317703
Formula:
C₁₄H₁₈BrNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD01317703 CAS Number: 261360-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...