N-Boc-3-methyl-L-phenylalanine 98%
Supplier: Thermo Scientific
Synonyms:
Boc-Phe(3-Me)-OH
H62860.03
H62860.06
H62860.03EA
144
EUR
H62860.03
H62860.06
N-Boc-3-methyl-L-phenylalanine 98%
N-Boc-3-methyl-L-phenylalanine
Purity: 98%
CAS No.: 114873-06-2
Molecular Formula: C15H21NO4
Formula Weight: 279.33
Physical Form: Solid
Beilstein Reference: 8989098
MDL No.: MFCD01862944
Formula:
C₁₅H₂₁NO₄ MW: 279,336 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01862944 CAS Number: 114873-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...