5-Methylthiazole-2-carboxylic acid 97%
Supplier: Thermo Scientific
|
Warning
|
H63076.03
H63076.06
H63076.03EA
0
EUR
H63076.03
H63076.06
5-Methylthiazole-2-carboxylic acid 97%
5-Methylthiazole-2-carboxylic acid
Purity: 97%
CAS No.: 61291-21-2
Molecular Formula: C5H5NO2S
Formula Weight: 143.16
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD08060736
Formula:
C₅H₅NO₂S MW: 143,16 g/mol |
MDL Number:
MFCD08060736 CAS Number: 61291-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...