DL-m-Tyrosine 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
H63163.03
H63163.06
H63163.03EA
0
EUR
H63163.03
H63163.06
DL-m-Tyrosine 98%
DL-m-Tyrosine
Purity: 98%
CAS No.: 775-06-4
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Physical Form: Powder
Appearance: Off-white to yellow
Melting Point: 280-285° dec
Beilstein Reference: 2416853
MDL No.: MFCD00002597
Formula:
C₉H₁₁NO₃ Melting Pt: 280…285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002597 CAS Number: 775-06-4 EINECS: 212-270-9 Merck Index: 14,9840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...