2-Fluoro-3-nitrotoluene 98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Fluoro-1-methyl-3-nitrobenzene

H63173.14 H63173.22
H63173.14EA 104 EUR
H63173.14 H63173.22
2-Fluoro-3-nitrotoluene 98%
2-Fluoro-3-nitrotoluene

Purity: 98% CAS No.: 437-86-5 Molecular Formula: C7H6FNO2 Formula Weight: 155.13 Flash Point: 98°(208°F) Physical Form: Liquid Density: 1.276 Refractive Index: 1.5290 Appearance: Colourless to pale yellow Melting Point: 19° dec. Boiling Point: 111°/12mm Beilstein Reference: 2442082 UN No.: UN2810 MDL No.: MFCD03412242

Formula: C₇H₆FNO₂
MW: 155,13 g/mol
Boiling Pt: 111 °C (12 mmHg)
Melting Pt: 19 °C (dec.)
Density: 1,276
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD03412242
CAS Number: 437-86-5
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR