4-Hydroxy-1H-indole-6-carboxylic acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H63236.MD
H63236.03
H63236.MDEA
0
EUR
H63236.MD
H63236.03
4-Hydroxy-1H-indole-6-carboxylic acid 97%
4-Hydroxy-1H-indole-6-carboxylic acid
Purity: 97%
CAS No.: 40990-52-1
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Pale brown to brown
MDL No.: MFCD04973988
Formula:
C₉H₇NO₃ |
MDL Number:
MFCD04973988 CAS Number: 40990-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...