2-Chloro-4-fluoro-5-nitrotoluene 97%
Supplier: Thermo Scientific
|
Warning
|
H63244.03
H63244.06
H63244.03EA
0
EUR
H63244.03
H63244.06
2-Chloro-4-fluoro-5-nitrotoluene 97%
2-Chloro-4-fluoro-5-nitrotoluene
Purity: 97%
CAS No.: 112108-73-3
Molecular Formula: C7H5ClFNO2
Formula Weight: 189.57
Physical Form: Powder
Appearance: Yellow
UN No.: UN2811
MDL No.: MFCD11110549
Formula:
C₇H₅ClFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11110549 CAS Number: 112108-73-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...