4-(Bromomethyl)-3-fluorobenzonitrile 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 95%
CAS No.: 105942-09-4
Molecular Formula: C8H5BrFN
Formula Weight: 214.04
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Off-white
Melting Point: 75-79° dec
Beilstein Reference: 7758134
UN No.: UN2923
MDL No.: MFCD07368341
Formula:
C₈H₅BrFN Melting Pt: 75…79 °C (dec.) |
MDL Number:
MFCD07368341 CAS Number: 105942-09-4 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...