1-Methyl-L-glutamate 98%
Supplier: Thermo Fisher Scientific
Purity: 98%
CAS No.: 6384-08-3
Molecular Formula: C6H11NO4
Formula Weight: 161.16
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 145° dec.
Beilstein Reference: 1707273
Optical Rotation: +35°(c=2 in water)
MDL No.: MFCD00171677
Formula:
C₆H₁₁NO₄ Melting Pt: 145 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00171677 CAS Number: 6384-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...