(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%

Supplier: Thermo Fisher Scientific

H63342.06 H63342.14
H63342.06EA 102 EUR
H63342.06 H63342.14
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid

Purity: 98% CAS No.: 133174-15-9 Molecular Formula: C21H23N3O5 Formula Weight: 397.42 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 7780987 MDL No.: MFCD00151943

Formula: C₂₁H₂₃N₃O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00151943
CAS Number: 133174-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR