L-Tyrosine benzyl ester-p-toluenesulphonate 98%
Supplier: Thermo Scientific
Synonyms:
L-Tyrosine benzyl ester p-toluenesulphonate salt, H-Tyr-OBzl·TosOH, H-Tyr-OBzl.Tos-OH
H63361.14
H63361.14EA
0
EUR
H63361.14
L-Tyrosine benzyl ester-p-toluenesulphonate 98%
L-Tyrosine benzyl ester-p-toluenesulphonate
Purity: 98%
CAS No.: 53587-11-4
Molecular Formula: C16H17NO3.C7H8O3S
Formula Weight: 443.51
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 3855339
MDL No.: MFCD00038959
Formula:
C₁₆H₁₇NO₃ MW: 443,51 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00038959 CAS Number: 53587-11-4 EINECS: 258-650-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...