1-Benzylindole 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H63414.03
H63414.06
H63414.03EA
107
EUR
H63414.03
H63414.06
1-Benzylindole 97%
1-Benzylindole
Purity: 97%
CAS No.: 3377-71-7
Molecular Formula: C15H13N
Formula Weight: 207.27
Physical Form: Solid
Appearance: Off-white to yellow
Beilstein Reference: 148884
MDL No.: MFCD00015884
Formula:
C₁₅H₁₃N Storage Temperature: Ambient |
MDL Number:
MFCD00015884 CAS Number: 3377-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...