2-chloro-5-iodopyrimidine 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Chloro-5-iodopyrimidine
H63423.03
H63423.06
H63423.03EA
75.7
EUR
H63423.03
H63423.06
2-chloro-5-iodopyrimidine 95%
2-chloro-5-iodopyrimidine
Purity: 95%
CAS No.: 32779-38-7
Molecular Formula: C4H2ClIN2
Formula Weight: 240.43
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Off-white
MDL No.: MFCD08062390
Formula:
C₄H₂ClIN₂ MW: 240.428 g/mol |
MDL Number:
MFCD06200210 CAS Number: 32779-38-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...