3-Hydroxyisonicotinic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Hydroxypyridine-4-carboxylic acid
H63465.03
H63465.06
H63465.03EA
34.2
EUR
H63465.03
H63465.06
3-Hydroxyisonicotinic acid 98%
3-Hydroxyisonicotinic acid
Purity: 98%
CAS No.: 10128-71-9
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Physical Form: Solid
Beilstein Reference: 383677
MDL No.: MFCD00234165
Formula:
C₆H₅NO₃ MW: 139,11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234165 CAS Number: 10128-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...