3-(4-Thiazolyl)-L-alanine 95%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
H-3-(4-Thiazolyl)-Ala-OH, (S)-2-Amino-3-(thiazol-4-yl)propanoic acid
H63511.MD
H63511.03
H63511.MDEA
0
EUR
H63511.MD
H63511.03
3-(4-Thiazolyl)-L-alanine 95%
3-(4-Thiazolyl)-L-alanine
Purity: 95%
CAS No.: 119433-80-6
Molecular Formula: C6H8N2O2S
Formula Weight: 172.20
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Pale yellow
MDL No.: MFCD00079677
Formula:
C₆H₈N₂O₂S MW: 172.202 g/mol |
MDL Number:
MFCD00079677 CAS Number: 119433-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...